Assay Details
Gene Symbol: Abcg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004076.2 | NP_001004076.2 | 15 - 16 | 1867 | 112 | |
XM_008770002.1 | XP_008768224.1 | 15 - 16 | 4257 | 112 | ||
XM_008770003.1 | XP_008768225.1 | 16 - 17 | 4137 | 112 | ||
GenBank mRNA | AY688113.1 | - | 14 - 15 | 1794 | 112 | |
BC098896.1 | - | 15 - 16 | 1867 | 7100556 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P552655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113346 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P552655 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P552655 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P552655 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P552655 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71370 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4156 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104448 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56424 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11384 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10775296 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775327 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775330 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775329 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775336 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775332 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775297 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775294 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775337 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775331 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775293 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775333 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775300 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775299 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775334 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775335 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775328 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10775326 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10775283 | - | - |
Rat Genome 230 2.0 Array | 1393217_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393217_at | - | - |
Affymetrix HT Rat Focus Array | 1393217_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393217_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400009725.rn.1 | TC1400001304.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400009727.rn.1 | TC1400001304.rn.1 | RTA TC |
Rat Expression Set 230 | 1393217_at | - | - |
Rat Expression Set 230 | 1393217_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775326 | - | - |
Rat Genome U34 Set | rc_AI169162_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20919090 | - | - |
ABI Rat Genome Survey Microarray | 21343570 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376371 | - | - |
