Assay Details
Gene Symbol: Nkx6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031737.1 | NP_113925.1 | 1 - 2 | 1458 | 90 | |
GenBank mRNA | AF004431.1 | - | 1 - 2 | 1458 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
