Assay Details
| Gene Symbol: Hnrpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082539.1 | NP_001076008.1 | 3 - 4 | 842 | 95 | |
| NM_001082540.1 | NP_001076009.1 | 4 - 5 | 899 | 95 | ||
| NM_001082541.1 | NP_001076010.1 | 3 - 4 | 842 | 95 | ||
| NM_024404.2 | NP_077380.2 | 4 - 5 | 899 | 95 | ||
| GenBank mRNA | AB046615.1 | - | 4 - 5 | 663 | 95 | |
| AB046616.1 | - | 3 - 4 | 557 | 95 | ||
| AB046617.1 | - | 4 - 5 | 614 | 95 | ||
| AB046618.1 | - | 3 - 4 | 557 | 95 | ||
| FQ212501.1 | - | 1 - 2 | 152 | 95 | ||
| Gene Symbol: Hnrnpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X16933.1 | - | 176 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrpd
Genomic Map