Assay Details
| Gene Symbol: Hnrpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082539.1 | NP_001076008.1 | 4 - 5 | 964 | 93 | |
| NM_001082540.1 | NP_001076009.1 | 5 - 6 | 1021 | 93 | ||
| NM_001082541.1 | NP_001076010.1 | 4 - 5 | 964 | 93 | ||
| NM_024404.2 | NP_077380.2 | 5 - 6 | 1021 | 93 | ||
| GenBank mRNA | AB046615.1 | - | 5 - 6 | 785 | 93 | |
| AB046616.1 | - | 4 - 5 | 679 | 93 | ||
| AB046617.1 | - | 5 - 6 | 736 | 93 | ||
| AB046618.1 | - | 4 - 5 | 679 | 93 | ||
| FQ212501.1 | - | 2 - 3 | 274 | 93 | ||
| Gene Symbol: Hnrnpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X16933.1 | - | 298 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrpd
Genomic Map