Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025670.1 | NP_001020841.1 | 8 - 9 | 1211 | 136 | |
| XM_008770019.1 | XP_008768241.1 | 9 - 10 | 1208 | 136 | ||
| XM_008770020.1 | XP_008768242.1 | 9 - 10 | 1637 | 136 | ||
| XM_008770021.1 | XP_008768243.1 | 9 - 10 | 1336 | 136 | ||
| GenBank mRNA | BC097362.1 | - | 8 - 9 | 1211 | 7442656 | 136 |
More Information
Updated Mapping Information
Set Membership: |
|

Agpat9
Genomic Map