Assay Details
Gene Symbol: Hsd17b11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004209.1 | NP_001004209.1 | 1 - 2 | 266 | 59 | |
XM_006250627.2 | XP_006250689.1 | 2 - 3 | 453 | 59 | ||
XM_006250629.2 | XP_006250691.1 | 2 - 3 | 377 | 59 | ||
GenBank mRNA | BC078929.1 | - | 1 - 2 | 266 | 7107084 | 59 |
FQ210820.1 | - | 1 - 2 | 282 | 59 | ||
FQ222122.1 | - | 1 - 2 | 279 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
