Assay Details
| Gene Symbol: Bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107292.1 | NP_001100762.1 | 6 - 7 | 884 | 73 | |
| XM_006252606.2 | XP_006252668.1 | 6 - 7 | 563 | 73 | ||
| XM_006252607.2 | XP_006252669.1 | 6 - 7 | 563 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bap1
Genomic Map