Assay Details
Gene Symbol: Prl2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053791.2 | NP_446243.1 | 3 - 4 | 358 | 75 | |
GenBank mRNA | AF226608.1 | - | 3 - 4 | 356 | 75 | |
BC097296.1 | - | 3 - 4 | 358 | 7445776 | 75 | |
DQ329279.1 | - | 3 - 4 | 356 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
