Assay Details
Gene Symbol: Prl7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053364.1 | NP_445816.1 | 1 - 2 | 110 | 95 | |
GenBank mRNA | AF139809.1 | - | 1 - 2 | 110 | 95 | |
AF226609.1 | - | 1 - 2 | 82 | 95 | ||
FQ219949.1 | - | 1 - 2 | 106 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF139809.1, AF226609.1, NM_053364.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
