Assay Details
Gene Symbol: Prl7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053364.1 | NP_445816.1 | 2 - 3 | 285 | 129 | |
GenBank mRNA | AF139809.1 | - | 2 - 3 | 285 | 129 | |
AF226609.1 | - | 2 - 3 | 257 | 129 | ||
FQ219949.1 | - | 2 - 3 | 281 | 129 | ||
FQ220120.1 | - | 2 - 3 | 232 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
