Assay Details
| Gene Symbol: Gmds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039606.1 | NP_001034695.1 | 4 - 5 | 368 | 101 | |
| GenBank mRNA | BC104708.1 | - | 4 - 5 | 368 | 7317156 | 101 |
More Information
Updated Mapping Information
Set Membership: |
|

Gmds
Genomic Map