Assay Details
| Gene Symbol: Dusp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108510.1 | NP_001101980.1 | 1 - 2 | 72 | 93 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108510.1 transcripts.
Set Membership: |
|

Dusp8
Genomic Map