Assay Details
Gene Symbol: Ankrd13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108514.1 | NP_001101984.1 | 10 - 11 | 1221 | 68 | |
XM_006230842.2 | XP_006230904.1 | 10 - 11 | 1248 | 68 | ||
GenBank mRNA | FQ213072.1 | - | 5 - 6 | 490 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
