Assay Details
| Gene Symbol: Nudt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760189.1 | XP_008758411.1 | 3 - 4 | 614 | 76 | |
| XM_341975.7 | XP_341976.4 | 3 - 4 | 569 | 76 | ||
| GenBank mRNA | FQ217198.1 | - | 2 - 3 | 112 | 76 | |
More Information
Set Membership: |
|

Nudt8
Genomic Map