Assay Details
| Gene Symbol: Hrasls5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039007.1 | NP_001034096.1 | 3 - 4 | 876 | 102 | |
| XM_006230768.2 | XP_006230830.1 | 3 - 4 | 608 | 102 | ||
| GenBank mRNA | AB255646.1 | - | 3 - 4 | 401 | 102 | |
| BC099084.1 | - | 3 - 4 | 876 | 7452977 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hrasls5
Genomic Map