Assay Details
| Gene Symbol: Minpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019263.1 | NP_062136.1 | 2 - 3 | 813 | 74 | |
| XM_008760326.1 | XP_008758548.1 | 2 - 3 | 390 | 74 | ||
| GenBank mRNA | AF012714.2 | - | 2 - 3 | 813 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Minpp1
Genomic Map