Assay Details
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271366.1 | NP_001258295.1 | 6 - 7 | 1609 | 104 | |
XM_006230453.2 | XP_006230515.1 | 6 - 7 | 1604 | 104 | ||
GenBank mRNA | FQ230633.1 | - | 1 - 2 | 841 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
