Assay Details
| Gene Symbol: Ust5r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760213.1 | XP_008758435.1 | 7 - 8 | 2102 | 114 | |
| GenBank mRNA | BC081730.1 | - | 7 - 8 | 1808 | 114 | |
| BC098905.1 | - | 7 - 8 | 1465 | 7325342 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ust5r
Genomic Map