Assay Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108513.1 | NP_001101983.1 | 9 - 10 | 1458 | 71 | |
| XM_006230833.1 | XP_006230895.1 | 9 - 10 | 1425 | 71 | ||
| GenBank mRNA | BC088464.1 | - | 8 - 9 | 1665 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc93b1
Genomic Map