Assay Details
Gene Symbol: F9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031540.1 | NP_113728.1 | 2 - 3 | 230 | 228 | |
GenBank mRNA | FM036569.1 | - | 2 - 3 | 194 | 228 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
