Assay Details
| Gene Symbol: Magec2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025713.1 | NP_001020884.1 | 1 - 2 | 31 | 100 | |
| GenBank mRNA | BC097464.1 | - | 1 - 2 | 31 | 7457620 | 100 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC097464.1, NM_001025713.1 transcripts.
Set Membership: |
|

Magec2
Genomic Map