Assay Details
| Gene Symbol: LOC500124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024329.1 | NP_001019500.1 | 1 - 2 | 136 | 119 | |
| GenBank mRNA | BC078992.1 | - | 1 - 2 | 136 | 7112600 | 119 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC078992.1,NM_001024329.1 transcripts.
Set Membership: |
|

LOC500124
Genomic Map