Assay Details
| Gene Symbol: LOC500124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024329.1 | NP_001019500.1 | 3 - 4 | 471 | 100 | |
| GenBank mRNA | BC078992.1 | - | 3 - 4 | 471 | 7112600 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC500124
Genomic Map