Assay Details
Gene Symbol: Nt5c3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107862.1 | NP_001101332.1 | 9 - 10 | 975 | 118 | |
XM_006236550.2 | XP_006236612.1 | 9 - 10 | 2123 | 118 | ||
XM_006236552.2 | XP_006236614.1 | 8 - 9 | 1017 | 118 | ||
GenBank mRNA | BC166444.1 | - | 9 - 10 | 997 | 8363316 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P054097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41742 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054097 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054097 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P054097 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P816767 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58535 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103289 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10862736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862745 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862741 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862746 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862744 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862743 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10862735 | - | - |
Rat Genome 230 2.0 Array | 1373412_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373412_at | - | - |
Affymetrix HT Rat Focus Array | 1373412_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373412_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400021124.rn.1 | TC0400002970.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400021125.rn.1 | TC0400002970.rn.1 | RTA TC |
Rat Expression Set 230 | 1373412_PM_at | - | - |
Rat Expression Set 230 | 1373412_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10862735 | - | - |
Rat Genome U34 Set | rc_AA956916_at | - | - |
Rat Genome U34 Set | rc_AI170538_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20759408 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354867 | - | - |
