Assay Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006236447.2 | XP_006236509.1 | 14 - 15 | 3727 | 87 | |
| XM_006236448.2 | XP_006236510.1 | 14 - 15 | 3740 | 87 | ||
| XM_008762931.1 | XP_008761153.1 | 14 - 15 | 3724 | 87 | ||
| XM_342681.5 | XP_342682.2 | 14 - 15 | 3709 | 87 | ||
More Information
Set Membership: |
|

Krba1
Genomic Map