Assay Details
| Gene Symbol: Lamtor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106441.1 | NP_001099911.1 | 1 - 2 | 141 | 129 | |
| XM_008761134.1 | XP_008759356.1 | 1 - 2 | 167 | 129 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001106441.1,XM_008761134.1 transcripts.
This assay is in the 5'UTR region of the NM_001106441.1,XM_008761134.1 transcripts.
Set Membership: |
|

Lamtor2
Genomic Map