Assay Details
Gene Symbol: Bend6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108792.1 | NP_001102262.1 | 4 - 5 | 672 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of neuron differentiation
negative regulation of Notch signaling pathway
RNA polymerase II transcription corepressor activity
chromatin binding
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of neuron differentiation
negative regulation of Notch signaling pathway
RNA polymerase II transcription corepressor activity
chromatin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P606344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13224 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P606344 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P606344 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P606344 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P606344 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72083 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10927254 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10927257 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10927258 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10927253 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10927255 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10927256 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10927251 | - | - |
Affymetrix Rat Clariom S Assay | PSR0900003386.rn.1 | TC0900000434.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900003384.rn.1 | TC0900000434.rn.1 | RTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21704998 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351982 | - | - |
