Assay Details
Gene Symbol: Tsga10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030022.1 | NP_001025193.1 | 16 | 1817 | 80 | |
GenBank mRNA | AF092091.1 | - | 16 | 1817 | 80 | |
BC168202.1 | - | 14 - 15 | 1745 | 7115410 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
