Assay Details
Gene Symbol: Akr1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012498.1 | NP_036630.1 | 2 - 3 | 268 | 75 | |
GenBank mRNA | BC062034.1 | - | 2 - 3 | 253 | 5623700 | 75 |
X05884.1 | - | 2 - 3 | 265 | 75 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
