Assay Details
| Gene Symbol: Cdc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013103.2 | NP_001013121.2 | 2 - 3 | 281 | 140 | |
| GenBank mRNA | CB611861.1 | - | 2 - 3 | 284 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc34
Genomic Map