Assay Details
| Gene Symbol: Timm13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145781.2 | NP_665724.1 | 1 - 2 | 149 | 83 | |
| GenBank mRNA | AF144701.1 | - | 1 - 2 | 149 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Timm13
Genomic Map