Assay Details
Gene Symbol: Mrpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_573060.5 | XP_573060.3 | 2 - 3 | 513 | 92 | |
GenBank mRNA | FQ219872.1 | - | 2 - 3 | 485 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
