Assay Details
| Gene Symbol: Pisd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001061408.5 | XP_001061408.2 | 4 - 5 | 585 | 87 | |
| XM_006251423.2 | XP_006251485.1 | 4 - 5 | 4515 | 87 | ||
| XM_006251424.2 | XP_006251486.1 | 4 - 5 | 3697 | 87 | ||
| XM_006251426.2 | XP_006251488.1 | 3 - 4 | 4279 | 87 | ||
| XM_008770389.1 | XP_008768611.1 | 3 - 4 | 3824 | 87 | ||
More Information
Set Membership: |
|

Pisd
Genomic Map