Assay Details
| Gene Symbol: Tnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024771.1 | NP_001019942.1 | 2 - 3 | 668 | 115 | |
| XM_006251255.2 | XP_006251317.1 | 2 - 3 | 1015 | 115 | ||
| GenBank mRNA | BC085912.1 | - | 2 - 3 | 599 | 115 | |
| BC097956.1 | - | 2 - 3 | 668 | 7461734 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnip2
Genomic Map