Assay Details
| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107121.1 | NP_001100591.1 | 2 - 3 | 496 | 74 | |
| XM_008768963.1 | XP_008767185.1 | 2 - 3 | 2918 | 74 | ||
| XM_008768964.1 | XP_008767186.1 | 3 - 4 | 2569 | 74 | ||
| XM_008768965.1 | XP_008767187.1 | 2 - 3 | 2452 | 74 | ||
| XM_008768966.1 | XP_008767188.1 | 3 - 4 | 2509 | 74 | ||
| XM_008768967.1 | XP_008767189.1 | 2 - 3 | 2040 | 74 | ||
| XM_008768968.1 | XP_008767190.1 | 2 - 3 | 484 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp316
Genomic Map