Assay Details
Gene Symbol: Acy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009603.1 | NP_001009603.1 | 4 - 5 | 725 | 127 | |
XM_006230733.2 | XP_006230795.1 | 4 - 5 | 734 | 127 | ||
XM_006230734.2 | XP_006230796.1 | 4 - 5 | 787 | 127 | ||
XM_008760098.1 | XP_008758320.1 | 4 - 5 | 895 | 127 | ||
GenBank mRNA | BC088190.1 | - | 4 - 5 | 725 | 7371632 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
