Assay Details
| Gene Symbol: Eef1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004223.2 | NP_001004223.1 | 2 - 3 | 232 | 81 | |
| GenBank mRNA | BC079398.1 | - | 3 - 4 | 241 | 7112627 | 81 |
| BC098895.1 | - | 2 - 3 | 232 | 7367246 | 81 | |
| FQ212596.1 | - | 3 - 4 | 250 | 81 | ||
| FQ214210.1 | - | 3 - 4 | 248 | 81 | ||
| FQ214443.1 | - | 3 - 4 | 249 | 81 | ||
| FQ215853.1 | - | 3 - 4 | 278 | 81 | ||
| FQ219298.1 | - | 3 - 4 | 250 | 81 | ||
| FQ223614.1 | - | 3 - 4 | 248 | 81 | ||
| FQ224956.1 | - | 2 - 3 | 207 | 81 | ||
| FQ228703.1 | - | 3 - 4 | 248 | 81 | ||
| FQ228918.1 | - | 3 - 4 | 249 | 81 | ||
| FQ230995.1 | - | 3 - 4 | 248 | 81 | ||
| FQ231923.1 | - | 3 - 4 | 248 | 81 | ||
| FQ233868.1 | - | 3 - 4 | 248 | 81 | ||
| FQ234035.1 | - | 3 - 4 | 248 | 81 | ||
| FQ235034.1 | - | 3 - 4 | 398 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eef1g
Genomic Map