Assay Details
| Gene Symbol: Ndufv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006972.1 | NP_001006973.1 | 7 - 8 | 1092 | 59 | |
| GenBank mRNA | BC083709.1 | - | 7 - 8 | 1092 | 7190583 | 59 |
| FQ214688.1 | - | 7 - 8 | 1100 | 59 | ||
| FQ217076.1 | - | 7 - 8 | 1150 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufv1
Genomic Map