Assay Details
| Gene Symbol: LOC100911186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748886.2 | XP_003748934.1 | 6 - 7 | 779 | 101 | |
| Gene Symbol: Echs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078623.2 | NP_511178.1 | 6 - 7 | 751 | 101 | |
| GenBank mRNA | BC064655.1 | - | 6 - 7 | 751 | 5599860 | 101 |
| X15958.1 | - | 6 - 7 | 745 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911186
Genomic Map