Assay Details
| Gene Symbol: Dmwd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759036.1 | XP_008757258.1 | 3 - 4 | 1947 | 108 | |
| XM_008759037.1 | XP_008757259.1 | 3 - 4 | 1947 | 108 | ||
| GenBank mRNA | BC160851.1 | - | 3 - 4 | 1844 | 108 | |
More Information
Set Membership: |
|

Dmwd
Genomic Map