Assay Details
| Gene Symbol: Sh3d19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232783.2 | XP_006232845.1 | 7 - 8 | 1777 | 84 | |
| XM_006232784.2 | XP_006232846.1 | 8 - 9 | 2122 | 84 | ||
| XM_006232785.2 | XP_006232847.1 | 7 - 8 | 1779 | 84 | ||
| XM_215597.8 | XP_215597.6 | 7 - 8 | 1775 | 84 | ||
More Information
Set Membership: |
|

Sh3d19
Genomic Map