Assay Details
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019315.2 | NP_062188.2 | 1 - 2 | 1233 | 148 | |
GenBank mRNA | AF284345.1 | - | 1 - 2 | 930 | 148 | |
AF292389.1 | - | 1 - 2 | 1233 | 148 | ||
U69884.1 | - | 1 - 2 | 937 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
