Assay Details
| Gene Symbol: mrpl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007637.1 | NP_001007638.1 | 3 - 4 | 358 | 100 | |
| XM_006232604.2 | XP_006232666.1 | 3 - 4 | 650 | 100 | ||
| XM_006232605.2 | XP_006232667.1 | 3 - 4 | 653 | 100 | ||
| XM_006232607.2 | XP_006232669.1 | 3 - 4 | 540 | 100 | ||
| XM_006232608.2 | XP_006232670.1 | 3 - 4 | 570 | 100 | ||
| GenBank mRNA | BC082018.1 | - | 3 - 4 | 358 | 7132470 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

mrpl24
Genomic Map