Assay Details
| Gene Symbol: Tmem79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232637.2 | XP_006232699.1 | 1 - 2 | 228 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232637.2 transcripts.
Set Membership: |
|

Tmem79
Genomic Map