Assay Details
Gene Symbol: LOC680112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109392.1 | NP_001102862.1 | 1 - 2 | 236 | 96 | |
XM_006236383.2 | XP_006236445.1 | 2 - 3 | 208 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
