Assay Details
Gene Symbol: Ptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017066.2 | NP_058762.1 | 2 - 3 | 178 | 83 | |
XM_006236290.2 | XP_006236352.1 | 2 - 3 | 1562 | 83 | ||
XM_006236294.2 | XP_006236356.1 | 2 - 3 | 1158 | 83 | ||
GenBank mRNA | BC062013.1 | - | 2 - 3 | 178 | 5621277 | 83 |
FQ213442.1 | - | 2 - 3 | 182 | 83 | ||
FQ223195.1 | - | 2 - 3 | 330 | 83 | ||
M55601.1 | - | 2 - 3 | 379 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
