Assay Details
Gene Symbol: Ptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017066.2 | NP_058762.1 | 1 - 2 | 67 | 76 | |
GenBank mRNA | BC062013.1 | - | 1 - 2 | 67 | 5621277 | 76 |
FQ213442.1 | - | 1 - 2 | 71 | 76 | ||
FQ223195.1 | - | 1 - 2 | 219 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC062013.1, NM_017066.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
