Assay Details
| Gene Symbol: Alyref | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109602.1 | NP_001103072.1 | 2 - 3 | 389 | 109 | |
| XM_006247925.2 | XP_006247987.1 | 2 - 3 | 384 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alyref
Genomic Map