Assay Details
| Gene Symbol: Ccl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004202.2 | NP_001004202.1 | 2 - 3 | 127 | 99 | |
| GenBank mRNA | AY620903.1 | - | 2 - 3 | 137 | 99 | |
| BC079460.1 | - | 159 | 7121977 | 99 | ||
| EF121992.1 | - | 2 - 3 | 127 | 99 | ||
| EF121993.1 | - | 2 - 3 | 127 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl6
Genomic Map