Assay Details
| Gene Symbol: Zswim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163492.1 | NP_001156964.1 | 2 - 3 | 107 | 121 | |
| GenBank mRNA | BF522197.1 | - | 2 - 3 | 114 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zswim7
Genomic Map